Learn More
Tocris Bioscience™ Disulfide biotin azide
Descripción
Disulfide Biotin Azide is an azide-activated cleavable (disulphide) biotin probe. It allows the biotinylation of molecules containing alkyne moiety via copper-catalyzed click chemistry reactions.
Especificaciones
Especificaciones
| Nombre del producto químico o material | N-(24-Azido-12,20-dioxo-3,6,9-trioxa-16,17-dithia-13,21-diazatetracosyl)-5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamide |
| CAS | 2866429-93-6 |
| Cantidad | 5 mg |
| Objetivo | General Laboratory Research Reagent |
| Fórmula molecular | C27H48N8O7S3 |