Learn More
MedChemExpress Diprotin A TFA
Product Code. 16453066
Encompass_Preferred
Change view
Click to view available options
Cantidad:
25 mg
Unit Size:
25 miligramos
Product Code. Cantidad unitSize
16453066 25 mg 25 miligramos
1 options
This item is not returnable. View return policy
Product Code. 16453066 Supplier MedChemExpress Supplier No. HY111174A
This item is not returnable. View return policy
MedChemExpress Diprotin A TFA (Ile-Pro-Ile TFA) is an inhibitor of dipeptidyl peptidase IV (DPP-IV).
Specifications
Pureza 98.8%
CAS 209248-71-5
Contenido y almacenamiento Sealed storage, away from moisture∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Para utilizar con (aplicación) Metabolism-protein/nucleotide metabolism
Peso molecular 455.47 g/mol
Tipo de producto Peptides
Cantidad 25 mg
Condiciones de envío Room temperature
Solubilidad H2O : ≥ 77.5 mg/mL (170.15 mM)
Sinónimo Ile-Pro-Ile TFA