Learn More
MedChemExpress Diphenylterazine
Especificaciones
Especificaciones
| CAS | 344940-63-2 |
| Solubilidad | DMSO : 27.5 mg/mL (72.86 mM∣Need ultrasonic∣DMSO can inactivate Diphenylterazine's activity) ∣DMF : 11.11 mg/mL (29.44 mM∣Need ultrasonic∣DMSO can inactivate Diphenylterazine's activity) ∣EtOH+HCl : 1 mg/mL (2.65 mM∣ultrasonic and warming and adjust pH to 2 with 1M HCl and heat to 80°C∣DMSO can inactivate Diphenylterazine's activity) ∣H2O : < 0.1 mg/mL (insoluble∣DMSO can inactivate Diphenylterazine's activity) |
| Sinónimo | DTZ |
| Contenido y almacenamiento | -80°C, protect from light |
| Cantidad | 50 mg |
| Peso molecular | 377.44 g/mol |
| Tipo de producto | Dye Reagents |
| Grado de pureza o calidad | 95.0% |
| Condiciones de envío | Room temperature |