missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DIO3 Polyclonal Antibody
GREENER_CHOICE

Código de producto. 13231379
Change view
Click to view available options
Cantidad:
400 μL
Tamaño de la unidad:
100 microgramos
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
13231379 400 μL 100 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 13231379 Proveedor Invitrogen™ N.º de proveedor PA526537

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

DIO3 Polyclonal Antibody for Western Blot, IHC (P)

DIO3 belongs to the iodothyronine deiodinase family. It catalyzes the inactivation of thyroid hormone by inner ring deiodination of the prohormone thyroxine (T4) and the bioactive hormone 3,3',5-triiodothyronine (T3) to inactive metabolites, 3,3',5'-triiodothyronine (RT3) and 3,3'-diiodothyronine (T2), respectively. This enzyme is highly expressed in the pregnant uterus, placenta, fetal and neonatal tissues, suggesting that it plays an essential role in the regulation of thyroid hormone inactivation during embryological development. This protein contains a selenocysteine (Sec) residue, which is essential for efficient enzyme activity. The selenocysteine is encoded by the UGA codon, which normally signals translation termination. The 3' UTR of Sec-containing genes have a common stem-loop structure, the sec insertion sequence (SECIS), which is necessary for the recognition of UGA as a Sec codon rather than as a stop signal.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno DIO3
Aplicaciones Immunohistochemistry (Paraffin), Western Blot
Clasificación Polyclonal
Concentración 0.5 mg/mL
Conjugado Unconjugated
Formulación PBS with 0.09% sodium azide; pH 7.4
génica DIO3
N.º de referencia del gen P49897, P55073
Alias de gen 5DIII; D3; deiodinase, iodothyronine type III; deiodinase, iodothyronine, type 3; deiodinase, iodothyronine, type III; Dio3; DIOIII; I79_015795; iodothyronine deiodinase 3; iodothyronine deiodinase, placental type; Itdi3; LOW QUALITY PROTEIN: thyroxine 5-deiodinase; thyroxine 5-deiodinase; thyroxine deiodinase; thyroxine deiodinase type III (selenoprotein); TXDI3; Type 3 DI; type 3 iodothyronine selenodeiodinase; type III iodothyronine deiodinase; type-III 5' deiodinase; type-III 5'-deiodinase
Símbolos de los genes DIO3
Especie del huésped Rabbit
Inmunógeno KLH conjugated synthetic peptide between 250-278 amino acids from the C-terminal region of human DIO3.
Método de purificación Antigen Affinity Chromatography, Protein A
Cantidad 400 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 1735, 29475
Especies diana Human, Rat
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.