Learn More
Diacetato de dihidrofluoresceína, 97 %, Thermo Scientific Chemicals
Descripción
Fluorescent dye for detecting intracellular oxidants
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Especificaciones
| Nombre del producto químico o material | Dihydrofluorescein diacetate |
| Punto de fusión | 213°C to 215°C |
| Color | Blanco a amarillo |
| Cantidad | 1 g |
| Beilstein | 364167 |
| Formula Weight (peso de la fórmula) | 418.39 |
| Porcentaje de pureza | 97% |
| Forma física | Polvo |