Learn More
Tocris Bioscience™ DiFMUP
Descripción
DiFMUP is a fluorescent phophatase substrate widely used for the detection of phosphatase activity. The reaction product of DiFMUP has excitation/emission maxima of 358/455 nm.
Note: This product is typically prepared in DMSO.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 6,8-Difluoro-4-methyl-7-(phosphonooxy)-2H-1-benzopyran-2-one |
| CAS | 214491-43-7 |
| Cantidad | 5 mg |
| Objetivo | Enzyme Probes and Enzyme Substrates |
| Fórmula molecular | C10H7F2O6P |
| Formula Weight (peso de la fórmula) | 292.13 |
| Tipo de producto | Fluorescent substrate |
| Pureza | 95% |