Learn More
Desmetil ranolazina-d5, TRC
Product Code. 30352887
Change view
Click to view available options
Cantidad:
10 mg
Unit Size:
10 miligramos
Product Code. Cantidad unitSize
30352887 10 mg 10 miligramos
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30352887 Supplier Toronto Research Chemicals Supplier No. TRCD29215210MG
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Moléculas orgánicas de alta pureza y estándares analíticos, entregados estratégicamente en todo el mundo para potenciar la innovación y el éxito comercial.
TRUSTED_SUSTAINABILITY
Specifications
Nombre del producto químico o material Desmethyl Ranolazine-d5
CAS 1329834-18-5
Fórmula molecular C23H26D5N3O4
Cantidad 10 mg
Sinónimo N-(2,6-Dimethylphenyl)-4-[2-hydroxy-3-(2-hydroxyphenoxy)propyl-d5]- 1-piperazineacetamide, RS 88390-d5
Fórmula InChI InChI=1S/C23H31N3O4/c1-17-6-5-7-18(2)23(17)24-22(29)15-26-12-10-25(11-13-26)14-19(27)16-30-21-9-4-3-8-20(21)28/h3-9,19,27-28H,10-16H2,1-2H3,(H,24,29)/i14D2,16D2,19D
SMILES OC1=CC=CC=C1OC([2H])([2H])C(O)([2H])C([2H])([2H])N2CCN(CC(NC3=C(C)C=CC=C3C)=O)CC2
Peso molecular (g/mol) 418.54