Learn More
Deschloroclozapine, MedChemExpress
Código de producto. 16488048
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
10 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
50 miligramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
16488048 10 mg 10 miligramos
16477826 100 mg 100 miligramos
16473967 50 mg 50 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 16488048 Proveedor Medchem Express N.º de proveedor HY42110
MedChemExpress Deschloroclozapine, a metabolite of Clozapine, is a highly potent muscarinic DREADDs agonist. Deschloroclozapine binds to DREADD receptor subtypes hM3Dq and hM4Di with Ki of 6.3 and 4.2 nM, respectively. [11C]-Deschloroclozapine is developed as a promising PET tracer for DREADD imaging.
Deschloroclozapine, a metabolite of Clozapine, is a highly potent muscarinic DREADDs agonist. Deschloroclozapine binds to DREADD receptor subtypes hM3Dq and hM4Di with Ki of 6.3 and 4.2 nM, respectively. [11C]-Deschloroclozapine is developed as a promising PET tracer for DREADD imaging.
Especificaciones
Nombre del producto químico o material Deschloroclozapine
Forma física Powder
CAS 7-7-1977
Color Amarillo claro
Cantidad 10 mg
Notas de grado de pureza Research
Fórmula molecular C18H20N4
Para utilizar con (aplicación) Neuroscience-Neuromodulation
Información de solubilidad DMSO : 100 mg/mL (342.02 mM; Need ultrasonic)
SMILES CN1CCN(C2=NC3=CC=CC=C3NC4=CC=CC=C42)CC1
Peso molecular (g/mol) 292.38
Formula Weight (peso de la fórmula) 292.38
Porcentaje de pureza 99.59%