Learn More
Delavirdine mesylate, MedChemExpress
Código de producto. 16470407
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
10 mg
50 mg
100 mg
200 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
200 miligramos
50 miligramos
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
16470407 50 mg 50 miligramos
16455036 100 mg 100 miligramos
16414058 10 mg 10 miligramos
16450996 200 mg 200 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Código de producto. 16470407 Proveedor Medchem Express N.º de proveedor HY10571A
MedChemExpress Delavirdine (U 90152) mesylate is a potent, highly specific and orally active non-nucleoside reverse transcriptase inhibitor (NNRTI). Delavirdine mesylate selectively inhibits HIV-1 reverse transcriptase (RT) (IC50=0.26 μM) over DNA polymerase α (IC50=440 μM) and polymerase δ (IC50>550 μM). Delavirdine mesylate is an inhibitor of HIV-1 replication and can can be used for the study of AIDs.
Delavirdine (U 90152) mesylate is a potent, highly specific and orally active non-nucleoside reverse transcriptase inhibitor (NNRTI). Delavirdine mesylate selectively inhibits HIV-1 reverse transcriptase (RT) (IC50=0.26 μM) over DNA polymerase α (IC50=440 μM) and polymerase δ (IC50>550 μM). Delavirdine mesylate is an inhibitor of HIV-1 replication and can can be used for the study of AIDs.
Especificaciones
Nombre del producto químico o material Delavirdine mesylate
Forma física Solid
CAS 147221-93-0
Color Off-White
Cantidad 50 mg
Notas de grado de pureza Research
Fórmula molecular C23H32N6O6S2
Para utilizar con (aplicación) COVID-19-anti-virus
Sinónimo U 90152 mesylateBHAP-U 90152 mesylate
Información de solubilidad DMSO : ≥ 40.3 mg/mL (72.92 mM)
SMILES O=S(NC1=CC=C(NC(C(N2CCN(C3=C(NC(C)C)C=CC=N3)CC2)=O)=C4)C4=C1)(C)=O.O=S(O)(C)=O
Peso molecular (g/mol) 552.67
Formula Weight (peso de la fórmula) 552.67
Porcentaje de pureza 97.33%