Learn More
Clorhidrato de deshidroaripiprazol-d8, TRC
Código de producto. 30356849
Change view
Click to view available options
Cantidad:
1 mg
Unit Size:
1 miligramo
Product Code. Cantidad unitSize
30356849 1 mg 1 miligramo
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30356849 Supplier Toronto Research Chemicals Supplier No. TRCD2291521MG
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Moléculas orgánicas de alta pureza y estándares analíticos, entregados estratégicamente en todo el mundo para potenciar la innovación y el éxito comercial.
TRUSTED_SUSTAINABILITY
Specifications
Nombre del producto químico o material Dehydroaripiprazole piperazinyl-2,2,3,3,5,5,6,6-d8 hydrochloride
CAS 1215383-78-0
Notas de grado de pureza HPLC
Fórmula molecular C23 2H8 H17 Cl2 N3 O2 . Cl H
Cantidad 1 mg
Sinónimo 7-[4-[2,2,3,3,5,5,6,6-Octadeuterio-4-(2,3-dichlorophenyl)piperazin-1-yl]butoxy]-1H-quinolin-2-one hydrochloride, Dehydro Aripiprazole-d8 hydrochloride, 7-[4-[4-(2,3-Dichlorophenyl) 2,2,3,3,5,5,6,6-d8-piperazin-1-yl]butoxy]quinolin-2(1H)-one hydrochloride, Dehydroaripiprazole-D8 hydrochloride (piperazinyl-2,2,3,3,5,5,6,6-D8), 7-[4-[4-(2,3-Dichlorophenyl)-1-piperazinyl-2,2,3,3,5,5,6,6-d8]butoxy]-2(1H)-quinolinone hydrochloride
Fórmula InChI InChI=1S/C23H25Cl2N3O2.ClH/c24-19-4-3-5-21(23(19)25)28-13-11-27(12-14-28)10-1-2-15-30-18-8-6-17-7-9-22(29)26-20(17)16-18;/h3-9,16H,1-2,10-15H2,(H,26,29);1H/i11D2,12D2,13D2,14D2;
SMILES Cl.[2H]C1([2H])N(CCCCOc2ccc3C=CC(=O)Nc3c2)C([2H])([2H])C([2H])([2H])N(c4cccc(Cl)c4Cl)C1([2H])[2H]
Nombre IUPAC 7-[4-[2,2,3,3,5,5,6,6-octadeuterio-4-(2,3-diclorofenil)piperazin-1-il]butoxi]-1H-quinolin-2-ona;clorhidrato
Peso molecular (g/mol) 490.88
Formula Weight (peso de la fórmula) 489.1593
Porcentaje de pureza >95