Learn More
Tocris Bioscience™ DC 260126
FFA1 (GPR40) antagonist
Marca: Tocris Bioscience™ 5357/10
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
DC 260126 is a free fatty acid receptor 1 (FFA1/GPR40) antagonist. Inhibits FFA-induced increases in intracellular Ca2+ levels and suppresses palmitic acid potentiated glucose-stimulated insulin secretion in Min6 pancreatic β cells in vitro. Decreases serum insulin levels, improves insulin sensitivity and reduces the number of apoptotic pancreatic β cells in obese diabetic (db/db) rats.Especificaciones
| N-(4-Butylphenyl)-4-fluorobenzenesulfonamide | |
| 346692-04-4 | |
| 10 mg |