Learn More
Tocris Bioscience™ DC 260126
Descripción
DC 260126 is a free fatty acid receptor 1 (FFA1/GPR40) antagonist. Inhibits FFA-induced increases in intracellular Ca2+ levels and suppresses palmitic acid potentiated glucose-stimulated insulin secretion in Min6 pancreatic β cells in vitro. Decreases serum insulin levels, improves insulin sensitivity and reduces the number of apoptotic pancreatic β cells in obese diabetic (db/db) rats.
Especificaciones
Especificaciones
| Nombre del producto químico o material | N-(4-Butylphenyl)-4-fluorobenzenesulfonamide |
| CAS | 346692-04-4 |
| Cantidad | 10 mg |
| Objetivo | Free Fatty Acid Receptor Antagonists |
| Fórmula molecular | C16H18FNO2S |
| Formula Weight (peso de la fórmula) | 307.38 |
| Pureza | >98% |