Learn More
Tocris Bioscience™ DBM 1285 dihydrochloride
GREENER_CHOICE
p38 MAPK inhibitor; anti-inflammatory
Marca:  Tocris Bioscience™ 5095/10
Encompass
Código de producto. 15706625
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos »
Descripción
Descripción
DBM 1285 dihydrochloride is a p38 MAPK inhibitor. Supresses p38 phosphorylation and LPS-induced TNF-α production in macrophages and in vivo. Attenuates zymosan-induced inflammation and adjuvant-induced arthritis in murine models. Orally active.
Especificaciones
Especificaciones
>98%
10 mg
Sugerencias de productos
Sugerencias de productos
Videos
SDS
Documentos
Documentos
Certificados
Promociones
Promociones