Learn More
Dasatinib, MedChemExpress
Código de producto. 16494686
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
100 mg
200 mg
500 mg
Tamaño de la unidad:
100 miligramos
200 miligramos
500 miligramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
16494686 100 mg 100 miligramos
16480936 200 mg 200 miligramos
16461536 500 mg 500 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 16494686 Proveedor Medchem Express N.º de proveedor HY10181
MedChemExpress Dasatinib (BMS-354825) is a highly potent, ATP competitive, orally active dual Src/Bcr-Abl inhibitor with potent antitumor activity. The Kis are 16 pM and 30 pM for Src and Bcr-Abl, respectively. Dasatinib inhibits Bcr-Abl and Src with IC50s of apoptosis and autophagy.
Dasatinib (BMS-354825) is a highly potent, ATP competitive, orally active dual Src/Bcr-Abl inhibitor with potent antitumor activity. The Kis are 16 pM and 30 pM for Src and Bcr-Abl, respectively. Dasatinib inhibits Bcr-Abl and Src with IC50s of apoptosis and autophagy.
Especificaciones
Nombre del producto químico o material Dasatinib
Forma física Solid
CAS 302962-49-8
Color Blanco
Cantidad 100 mg
Notas de grado de pureza Research
Fórmula molecular C22H26ClN7O2S
Para utilizar con (aplicación) Cancer-Kinase/protease
Sinónimo BMS-354825
Información de solubilidad DMSO : 250 mg/mL (512.28 mM; Need ultrasonic)
SMILES O=C(C1=CN=C(S1)NC2=NC(C)=NC(N3CCN(CC3)CCO)=C2)NC4=C(C=CC=C4Cl)C
Peso molecular (g/mol) 488.01
Formula Weight (peso de la fórmula) 488.01
Porcentaje de pureza 98.56%