Learn More
Tocris Bioscience™ DAPT
Descripción
DAPT is an ancillary Material Grade version of DAPT (Cat. No. 2634). Suitable for use as an ancillary reagent in the further manufacturing of cell therapies.
Find out more about our Ancillary Material Grade product range.
Especificaciones
Especificaciones
| Nombre del producto químico o material | (2 S)-N-[(3,5-Difluorophenyl)acetyl]-L-alanyl-2-phenylglycine 1,1-dimethylethyl ester |
| CAS | 208255-80-5 |
| Cantidad | 10 mg |
| Objetivo | Secretase Inhibitors |
| Fórmula molecular | C23H26F2N2O4 |
| Pureza | 0.99 |