Learn More
D609, MedChemExpress
Código de producto. 16494087
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
50 mg
Tamaño de la unidad:
10 miligramos
5 miligramos
50 miligramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
16494087 50 mg 50 miligramos
16488188 10 mg 10 miligramos
16400638 5 mg 5 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 16494087 Proveedor Medchem Express N.º de proveedor HY70072
MedChemExpress D609, an antitumoural xanthate, is a specific and competitive phosphatidyl choline-specific phospholipase C (PC-PLC) inhibitor with a Ki of 6.4 μM. D609 is an antioxidative protector and has antiviral and anti-inflammatory activity.
D609, an antitumoural xanthate, is a specific and competitive phosphatidyl choline-specific phospholipase C (PC-PLC) inhibitor with a Ki of 6.4 μM. D609 is an antioxidative protector and has antiviral and anti-inflammatory activity.
Especificaciones
Nombre del producto químico o material D609
Forma física Solid
CAS 83373-60-8
Color Blanco
Cantidad 50 mg
Notas de grado de pureza Research
Fórmula molecular C11H15KOS2
Para utilizar con (aplicación) Cancer-programmed cell death
Información de solubilidad DMSO : 100 mg/mL (375.29 mM; Need ultrasonic) ∣H2O : 2 mg/mL (7.51 mM; Need ultrasonic)
SMILES S=C(OC1C2C3CCCC3C(C1)C2)S[K]
Peso molecular (g/mol) 266.46
Formula Weight (peso de la fórmula) 266.46
Porcentaje de pureza 98.0%