Learn More
Tocris Bioscience™ CZC 25146, Tocris Bioscience™
Potent LRRK2 inhibitor
Marca: Tocris Bioscience™ 6071/50
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
CZC 25146 is a potent LRRK2 inhibitor (IC50 values are 4.76 and 6.87 nM for for wild-type and G2019S mutant forms of LRRK2, respectively). Also inhibits PLK4, GAK, TNK1, CAMKK2 and PIP4K2. Attenuates mutant LRRK2-induced injury of cultured rodent and human neurons. Cell permeable.Especificaciones
| CZC 25146 | |
| N-[2-[[5-Fluoro-2-[[2-methoxy-4-(4-morpholinyl)phenyl]amino]-4-pyrimidinyl]amino]phenyl]methanesulfonamide | |
| Soluble to 100mM in DMSO | |
| Pale Purple |
| 1191911-26-8 | |
| 50 mg | |
| Solid | |
| Bioactive Small Molecule |
Título del producto
For Research Use Only.