Learn More
Invitrogen™ Cytokeratin 14 Monoclonal Antibody (LL002)
GREENER_CHOICE
Código de producto. 11334483
Change view
Click to view available options
Cantidad:
500 μL
Tamaño de la unidad:
500 microlitros
11334483 500 μL 500 microlitros
1 options
Código de producto. 11334483 Proveedor Invitrogen™ N.º de proveedor MA511599
Mouse Monoclonal Antibody
MA5-11599 targets Cytokeratin 14 in IHC (P) and WB applications and shows reactivity with Human and Rat samples. The MA5-11599 immunogen is a synthetic peptide of 15 amino acid residues from the C-terminus of human keratin 14.
Cytokeratins (CK) are intermediate filaments of epithelial cells, both in keratinizing tissue (ie. skin) and non-keratinizing cells (ie. mesothelial cells). At least 20 different CKs can be identified. Biochemically, most members of the CK family fall into one of two classes, type I (acidic polypeptides) and type II (basic polypeptides). Belonging to the type I subfamily of low molecular weight keratins and existing in combination with keratin 5, keratin 14 distinguishes stratified epithelial cells from simple epithelial cells and is useful in identification of squamous cell carcinomas. It is considered a prognostic marker in breast carcinomas. At least one member of the acidic family and one member of the basic family is expressed in all epithelial cells.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Cytokeratin 14
Aplicaciones Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Multiplex Immunohistochemistry
Clasificación Monoclonal
Clon LL002
Concentración 0.2 mg/mL
Conjugado Unconjugated
Formulación PBS with 0.05% BSA and 0.05% sodium azide; pH 7.4
génica KRT14
N.º de referencia del gen P02533, Q61781, Q6IFV1
Alias de gen AI626930; CK14; CK-14; cytokeratin 14; cytokeratin-14; EBS3; EBS4; epidermolysis bullosa simplex, Dowling-Meara, Koebner; K14; Ka14; keratin 14; keratin 14 (epidermolysis bullosa simplex, Dowling-Meara, Koebner); keratin 14, type I; keratin complex 1, acidic, gene 14; keratin, type I cytoskeletal 14; keratin, type I cytoskeletal 17; keratin-14; Koebner; Krt-1.14; Krt1-14; KRT14; LOC100737113; NFJ; Type I keratin Ka14
Símbolos de los genes KRT14
Especie del huésped Mouse
Inmunógeno A synthetic peptide of 15 amino acid residues from the C-terminus of human keratin 14.
Método de purificación Protein A/G
Cantidad 500 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 16664, 287701, 3861
Especies diana Human, Mouse, Rat
Contenido y almacenamiento 4°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG3 κ