Learn More
Tocris Bioscience™ Cysmethynil
Descripción
Cysmethynil is an isoprenylcysteine carboxylmethyltransferase (ICMT) inhibitor (IC50 = 2.4 μM). Cysmethynil reduces growth of RAS-mutant cell lines in vitro and in vivo (EC50 = 20 μM) and reduces cell viability in a variety of cell lines (IC50 = 16.8-23.3 μM). Tumor growth, mTOR signaling and cell proliferation are inhibited by cysmethynil; apoptosis is induced. Cysmethynil increases p27 and p21 proteins and arrests cell-cycle at the G1 phase.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 5-(3-Methylphenyl)-1-octyl-1 H-indole-3-acetamide |
| CAS | 851636-83-4 |
| Cantidad | 10 mg |
| Objetivo | Other Transferase Inhibitors |
| Fórmula molecular | C25H32N2O |
| Pureza | 0.98 |