Learn More
Tocris Bioscience™ Cyprodime hydrochloride
Descripción
Selective μ-opioid receptor antagonist (Ki values are 5.4, 244.6 and 2187 nM for μ-, δ- and κ-opioid receptors respectively). Reduces levodopa-induced dyskinesia in the MPTP-lesioned primate model of Parkinson's disease.
Especificaciones
Especificaciones
| Nombre del producto químico o material | Cyprodime hydrochloride |
| CAS | 2387505-50-0 |
| Cantidad | 10 mg |
| Fórmula molecular | C22H30ClNO3 |
| Formula Weight (peso de la fórmula) | 391.93 |
| Pureza | >98% |
| Peso molecular | 391.94 |