missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CYP2A6 Monoclonal Antibody (OTI1D2)

Descripción
CYP2A6 Monoclonal Antibody for Western Blot, IHC (P), Flow
This gene, CYP2A6, encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and its expression is induced by phenobarbital. The enzyme is known to hydroxylate coumarin, and also metabolizes nicotine, aflatoxin B1, nitrosamines, and some pharmaceuticals. Individuals with certain allelic variants are said to have a poor metabolizer phenotype, meaning they do not efficiently metabolize coumarin or nicotine. This gene is part of a large cluster of cytochrome P450 genes from the CYP2A, CYP2B and CYP2F subfamilies on chromosome 19q. The gene was formerly referred to as CYP2A3; however, it has been renamed CYP2A6.
Especificaciones
Especificaciones
| Antígeno | CYP2A6 |
| Aplicaciones | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Clasificación | Monoclonal |
| Clon | OTI1D2 |
| Concentración | 1.0 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| génica | CYP2A6 |
| N.º de referencia del gen | P11509 |
| Alias de gen | 1,4-cineole 2-exo-monooxygenase; Coumarin 7-hydroxylase; CPA6; CYP; CYP2A; CYP2A3; CYP2A6; CYPIIA6; cytochrome P450 2A6; cytochrome P450 family 2 subfamily A member 6; cytochrome P450 IIA3; Cytochrome P450(I); cytochrome P450, family 2, subfamily A, polypeptide 6; cytochrome P450, subfamily IIA (phenobarbital-inducible), polypeptide 6; flavoprotein-linked monooxygenase; P450; P450 IIA3; P450(I); P450C2A; P450PB; xenobiotic monooxygenase |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?