Learn More
R&D Systems™ Cyno Monkey Matriptase/ST14 (XP_005580237.1) VersaClone cDNA
Especificaciones
Especificaciones
| génica | Matriptase/ST14 |
| Número de acceso | XP_005580237.1 |
| ID de gen (Entrez) | 6768 |
| Alias de gen | EC 3.4.21, Epithin, HAI, Matriptase, Membrane-type serine protease 1, MTSP1, MT-SP1EC 3.4.21.109, prostamin, PRSS14, Serine protease 14, Serine protease TADG-15, SNC19, SNC19MTSP1, ST14, suppression of tumorigenicity 14 (colon carcinoma), suppression of tumorigenicity 14 (colon carcinoma, matriptase, epithin), suppressor of tumorigenicity 14 protein, TADG15, TADG-15, TMPRSS14, tumor associated differentially expressed gene 15 protein, Tumor-associated differentially-expressed gene 15 protein |
| Contenido y almacenamiento | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Especie | Cynomolgus Monkey |
| Cantidad | 10 μg |