Learn More
R&D Systems™ Cyno Monkey E-Selectin (XP_005539998.1) VersaClone cDNA
Especificaciones
Especificaciones
| génica | E-Selectin/CD62E |
| Número de acceso | XP_005539998.1 |
| ID de gen (Entrez) | 6401 |
| Alias de gen | CD62 antigen-like family member E, CD62E, CD62E antigen, ELAM, ELAM1, ELAM-1, ELAM1E-selectin, endothelial adhesion molecule 1, Endothelial leukocyte adhesion molecule 1, ESEL, LECAM2, leukocyte endothelial cell adhesion molecule 2, Leukocyte-endothelial cell adhesion molecule 2, SELE, selectin E |
| Contenido y almacenamiento | Store the unopened product at -20 to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Especie | Cynomolgus Monkey |
| Línea de productos | R&D Systems |
| Cantidad | 10 μg |