Learn More
Clorhidrato de ciclopentamina, TRC
Especificaciones
Especificaciones
| Nombre del producto químico o material | Cyclopentamine Hydrochloride |
| CAS | 538-02-3 |
| Fórmula molecular | C9 H19 N . Cl H |
| Cantidad | 250 mg |
| Sinónimo | Cyclopentaneethanamine, N,α-dimethyl-, hydrochloride, Cyclopentamine hydrochloride, Cyclopentaneethanamine, N,α-dimethyl-, hydrochloride, Cyclopentaneethylamine, N,α-dimethyl-, hydrochloride, (±)-Cyclopentamine hydrochloride, 1-Cyclopentyl-2-methylaminopropane hydrochloride, Clopane hydrochloride, N,α-Dimethylcyclopentaneethylamine hydrochloride |
| Fórmula InChI | InChI=1S/C9H19N.ClH/c1-8(10-2)7-9-5-3-4-6-9;/h8-10H,3-7H2,1-2H3;1H |
| SMILES | Cl.CNC(C)CC1CCCC1 |
| Nombre IUPAC | 1-ciclopentil-N-metilpropan-2-amina;clorhidrato |
| Peso molecular (g/mol) | 177.71 |
| Formula Weight (peso de la fórmula) | 177.1284 |