missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Immunogen sequence: KAGSSKHHGP ISTTPGIIPQ KMSLDKYREK RKLETLDLDV RDHYIAAQVE QQHKQGQSQA ASSSSVTSPI KMKIPIANTE KYMADKKEKS GSLKLRIPIP PTDKSASKEE LKMKIKVSSS ERHSSSDEGS GKSKHSSPH Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 78%.
The protein encoded by this gene belongs to the highly conserved cyclin family, whose members are characterized by a dramatic periodicity in protein abundance through the cell cycle. Cyclins function as regulators of CDK kinases. Different cyclins exhibit distinct expression and degradation patterns which contribute to the temporal coordination of each mitotic event. This cyclin and its kinase partner CDK9 were found to be subunits of the transcription elongation factor p-TEFb. The p-TEFb complex containing this cyclin was reported to interact with, and act as a negative regulator of human immunodeficiency virus type 1 (HIV-1) Tat protein. A pseudogene of this gene is found on chromosome 1. Alternate splicing results in multiple transcript variants.
Especificaciones
Especificaciones
| Antígeno | Cyclin T2 |
| Aplicaciones | ChIP Assay, Immunohistochemistry (Paraffin), Immunocytochemistry |
| Clasificación | Polyclonal |
| Concentración | 0.2 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| génica | CCNT2 |
| N.º de referencia del gen | O60583 |
| Alias de gen | 2900041I18Rik; C81304; Ccnt2; cyclin T2; cyclin T2a; cyclin T2b; cyclin-T2; CycT2; CycT2a; CycT2b; SDS-stable vimentin-bound DNA fragment HEF42VIM22; subunit of positive elongation transcription factor b |
| Símbolos de los genes | CCNT2 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?
