missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cyclin B1 Monoclonal Antibody (5G6)
Descripción
MA5-15714 targets CCNB1 in indirect ELISA, FACS, IF, and WB applications and shows reactivity with Human and Rat samples. The MA5-15714 immunogen is purified recombinant fragment of human CCNB1 expressed in E. Coli. MA5-15714 detects CCNB1 which has a predicted molecular weight of approximately 60kDa.
Cyclins bind to and regulate the activity of the Cyclin dependent protein kinases (CDKs). Cyclin B1 or CCNB1 is a regulatory protein involved in mitosis. It is essential for the control of the cell cycle at the G2/M transition. Cyclin B1 complexes with p34 (cdc2) to form the maturation-promoting factor (MPF). Two alternative transcripts have been found, a constitutively expressed transcript and a cell cycle-regulated transcript, that is expressed predominantly during G2/M phase. Cyclin B1 is not ubiquitinated during G2/M phase, resulting in its steady accumulation during G2 phase, followed by abrupt APC dependent destruction at the end of mitosis. Destruction of Cyclin B1 is required for cell cycle progression. Cyclin B1 is overexpressed in various cancers, including breast, prostate, and non-small cell lung cancer.
Especificaciones
Especificaciones
| Antígeno | Cyclin B1 |
| Aplicaciones | ELISA, Flow Cytometry, Western Blot, Immunocytochemistry |
| Clasificación | Monoclonal |
| Clon | 5G6 |
| Concentración | Conc. Not Determined |
| Conjugado | Unconjugated |
| Formulación | Ascites with 0.03% sodium azide |
| génica | Ccnb1 |
| N.º de referencia del gen | P14635, P30277 |
| Alias de gen | Ccn-2; CCNB; Ccnb1; Ccnb1-rs1; Ccnb1-rs13; Cycb; Cycb1; Cycb1-rs1; Cycb-4; Cycb-5; cyclin B; cyclin B1; cyclin B1, related sequence 1; cyclin B1, related sequence 13; G2/mitotic-specific cyclin B1; G2/mitotic-specific cyclin-B1; I79_013589; OTTHUMP00000221981; OTTHUMP00000223023 |
| Mostrar más |
Título del producto
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?