Learn More
CYB5R1 Monoclonal Antibody (OTI2D10), TrueMAB™, OriGene
Código de producto. 17542102
Encompass_Preferred
Change view
Click to view available options
Cantidad:
30 μL
100 μL
Tamaño de la unidad:
100 microlitros
30 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
17542102 30 μL 30 microlitros
17522102 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 17542102 Proveedor Origene Technologies N.º de proveedor TA504639S
Mouse Monoclonal Antibody
CYB5R1 Monoclonal antibody specifically detects CYB5R1 in Canine, Human, Mouse, Rat samples. It is validated for Western Blot
Especificaciones
Antígeno CYB5R1
Aplicaciones Western Blot
Clasificación Monoclonal
Clon OTI2D10
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
génica Psmb7
N.º de referencia del gen P70195, Q99436, Q9JHW0
Alias de gen 1500005G05Rik; b5R.1; B5R1; B5R2; C80155; Cyb5r1; cytochrome b5 reductase 1; Humb5R2; NAD(P)H:quinone oxidoreductase type 3 polypeptide A2; NAD(P)H:quinone oxidoreductase type 3, polypeptide A2; NAD(P)H:quinone oxidoreductase type 3, polypeptide A2; cytochrome b5 reductase 1; NADH-cytochrome b5 reductase 1; Nqo3a2; UNQ3049/PRO9865
Especie del huésped Mouse
Inmunógeno Human recombinant protein fragment corresponding to amino acids 58-277 of human PSMB7 produced in E.coli.
Método de purificación Affinity chromatography
Cantidad 30 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 100686423, 19177, 5695, 85492
Especies diana Canine, Human, Mouse, Rat
Contenido y almacenamiento -20° C, Avoid Freeze/Thaw Cycles
Formulario Liquid
Isotype IgG1