Learn More
Tocris Bioscience™ Cyanine 5 Tyramide
Descripción
Cyanine 5 Tyramide is a red fluorescent reagent widely used for tyramide signal amplification (TSA) in IHC, ICC, FISH and multicolor FISH. HRP catalyzes localized deposition of multiple tyramide molecules (catalyzed reporter deposition, CARD), binding the fluorescein tyramide to adjacent tyrosines to enhance fluorescent signal.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 2-[5-[1,3-Dihydro-1-[6-[[(2-(4-hydroxyphenyl)ethyl]amino-6-oxohexyl]3,3-dimethyl-5-sulfo-2 H-indol-2-ylidene]-1,3-pentadien-1-yl]-1-ethyl-3,3-dimethyl-5-sulfo-3 H-indolium inner salt |
| CAS | 1431148-26-3 |
| Cantidad | 1 mg |
| Objetivo | Tyramide Signaling Amplification/TSA |
| Fórmula molecular | C41H49N3O8S2 |
| Formula Weight (peso de la fórmula) | 2020 |
| Pureza | 95% |