Learn More
Tocris Bioscience™ CX 614
Código de producto. 18765329
Change view
Click to view available options
Cantidad:
10 mg
50 mg
Tamaño de la unidad:
10 miligramos
50 miligramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
18765329 10 mg 10 miligramos
18716939 50 mg 50 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 18765329 Proveedor Tocris Bioscience™ N.º de proveedor 5149/10
AMPAkine; positive allosteric modulator of AMPA
CX 614 is an AMPAkine; positive allosteric modulator of AMPA. Prototypical benzamide which occupies subsite A only. Modulates channel deactivation and desensitization.
TRUSTED_SUSTAINABILITY
Especificaciones
Cantidad 10 mg
Tipo de producto CX 614
Pureza >99%
Peso molecular 247.25
Inhibidores CX 614
Para uso exclusivo en investigación