Learn More
Tocris Bioscience™ CS 640
Descripción
CS 640 is a potent calmodulin-dependent kinase I (CaMKI) inhibitor (IC50 values are 1 nM, 1 nM, 3 nM and 8 nM at CAMK1A, CAMK1G, CAMK1B and CAMK1D respectively). Improves insulin sensitivity and glucose control in a mouse model of diet-induced obesity. Orally bioavailable in rodents.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 2-[(3S)-3-Amino-1-piperidinyl]-4-[[2,6-bis(1-methylethyl)-4-pyridinyl]amino]-5-pyrimidinecarboxamide |
| CAS | 2388506-83-8 |
| Cantidad | 50 mg |
| Objetivo | CaM Kinase I |
| Fórmula molecular | C21H31N7O |
| Pureza | 0.98 |