Learn More
CRT 0066854 hydrochloride
PKCι and PKCζ inhibitor
Marca: Tocris Bioscience 5922/10
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
CRT 0066854 hydrochloride is a protein kinase C (PKC) inhibitor. Inhibits PKCι and PKCζ (IC50 values are 132 and 639 nM, respectively). Restores polarized morphogenesis in oncogene expressing MDCK cells, decreases colony formation in HeLa cells and inhibits lethal giant larvae 2 (LLGL2) phosphorylation.Especificaciones
| (2 S)-3-Phenyl-N1-[5,6,7,8-tetrahydro-2-(4-pyridinyl)[1 ]benzothieno[2,3-d]pyrimidin-4-yl]-1,2-propanediamine dihydrochloride | |
| 2250019-91-9 | |
| >98% | |
| 10 mg |