Learn More
CP 122721 hydrochloride, Tocris Bioscience™
High affinity NK1 receptor antagonist
Marca: Tocris Bioscience 3649/5
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
CP 122721 hydrochloride is a high affinity NK1 antagonist (pIC50 = 9.8). Suppresses recruitment of immunocytes in respiratory syncytial virus (RSV)-infected rats after capsaicin stimulation. Also blocks substance P-induced excitation of locus ceruleus cells in guinea pig brain slices (IC50 = 7 nM). Exhibits anxiolytic and antidepressant-like effects. Active in vivo.Especificaciones
| 145877-52-7 | |
| (2S,3S)-N-[[2-Methoxy-5-(trifluoromethoxy)phenyl]methyl]-2-phenyl-3-piperidinamine dihydrochloride | |
| 5 mg |