Learn More
Tocris Bioscience™ CP 122721 hydrochloride
Descripción
CP 122721 hydrochloride is a high affinity NK1 antagonist (pIC50 = 9.8). Suppresses recruitment of immunocytes in respiratory syncytial virus (RSV)-infected rats after capsaicin stimulation. Also blocks substance P-induced excitation of locus ceruleus cells in guinea pig brain slices (IC50 = 7 nM). Exhibits anxiolytic and antidepressant-like effects. Active in vivo.
Especificaciones
Especificaciones
| CAS | 145877-52-7 |
| Sinónimo | (2S,3S)-N-[[2-Methoxy-5-(trifluoromethoxy)phenyl]methyl]-2-phenyl-3-piperidinamine dihydrochloride |
| Cantidad | 25 mg |
| Fórmula molecular | C20H23F3N2O2.2HCl |
| Formula Weight (peso de la fórmula) | 453.33 |
| Pureza | 98% |