missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COX15 Monoclonal Antibody (2D2)

Descripción
Peptide Sequence: LTESGLSMVD WHLIKEMKPP TSQEEWEAEF QRYQQFPEFK ILNHDMTLTE FKFIWYMEYS H.
The cytochrome c oxidase (COX) family of proteins function as the final electron donor in the respiratory chain to drive a proton gradient across the inner mitochondrial membrane, ultimately resulting in the production of water. The mammalian COX apoenzyme is a dimer, with each monomer consisting of 13 subunits, some of which are mitochondrial and some of which are nuclear. COX15 (cytochrome c oxidase assembly protein COX15 homolog) is a 410 amino acid multi-pass mitochondrial membrane protein belonging to the COX15/ctaA family. Existing as two alternatively spliced isoforms, COX15 is predominantly expressed in tissues containing high rates of oxidative phosphorylation including brain, heart and muscle. COX15 defects are the cause of a neurodegenerative disorder known as Leigh syndrome and cytochrome c oxidase deficiency (COX deficiency).
Especificaciones
Especificaciones
| Antígeno | COX15 |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Monoclonal |
| Clon | 2D2 |
| Concentración | 0.2-1.0 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with no preservative; pH 7.4 |
| génica | COX15 |
| N.º de referencia del gen | Q7KZN9 |
| Alias de gen | 2900026G05Rik; CEMCOX2; COX15; COX15 cytochrome c oxidase assembly homolog; COX15 homolog, cytochrome c oxidase assembly protein; COX15, cytochrome c oxidase assembly homolog; cytochrome c oxidase assembly homolog 15; cytochrome c oxidase assembly homolog 15 (yeast); cytochrome c oxidase assembly protein 15; cytochrome c oxidase assembly protein COX15 homolog; cytochrome c oxidase subunit 15 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?