missing translation for 'onlineSavingsMsg'
Learn More

CoraLite™ Plus 488-conjugated GLUT1 Mouse Monoclonal Antibody, Proteintech

Código de producto. p-200108369 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
100 μL
20 μL
Tamaño de la unidad:
100 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
30236006 100 μL 100 microlitros
30234996 20 μL 20 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. View return policy
Product Code. 30236006 Supplier Proteintech Supplier No. CL48866290100UL

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The GLUT1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human GLUT1. This antibody recognizes human, mouse antigen. The GLUT1 antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™ plus 488.

Specifications

Antígeno GLUT1
Aplicaciones Flow Cytometry (Intracellular Staining)
Clasificación Monoclonal
Clon 2A5A2
Concentración 1000 μg/mL
Conjugado CoraLite Plus 488
Símbolos de los genes DYT17; DYT18; GLUT; GLUT1; MGC141895; MGC141896; PED
Especie del huésped Mouse
Inmunógeno Recombinant Protein
Método de purificación Protein G purification
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 6513
Especies diana Human, Mouse
Contenido y almacenamiento Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Peso molecular del anticuerpo 45-55 kDa
Tipo de producto Conjugated Antibody
Formulario Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.