Learn More
Carbonato de cobre(II) básico, puro, 54-56 % Cu, Thermo Scientific Chemicals
Código de producto. 10073903 Tienda Thermo Scientific Chemicals Productos
Change view
Click to view available options
Cantidad:
25 g
500 g
2.5 kg
Envase:
Frasco de vidrio
Tamaño de la unidad:
2.50 Kg
25 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad Envase unitSize
10073903 2.5 kg Frasco de vidrio 2.50 Kg
10713201 25 g Frasco de vidrio 25 g
10452391 500 g Frasco de vidrio 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 10073903 Proveedor Thermo Scientific Acros N.º de proveedor 196670025
Solicitar formato a granel o personalizado
CAS: 12069-69-1 | CCuO3·CuO2H2 | 221.1 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
TRUSTED_SUSTAINABILITY
CAS 12069-69-1
Fórmula molecular CCuO3·CuO2H2
Peso molecular (g/mol) 221.1
Número MDL MFCD00010976
Sinónimo Copper(II)hydroxide carbonate
Especificaciones
Nombre del producto químico o material Copper(II) carbonate basic
Punto de fusión 200.0°C
Color Verde
Forma física Polvo cristalino fino
Cantidad 2.5 kg
Fórmula lineal CuCO3·Cu(OH)2
Fieser 01,163
Índice Merck 15, 2621
Información de solubilidad Solubility in water: insoluble. Other solubilities: soluble in acids,ammonia,practically insoluble in alcohol
Formula Weight (peso de la fórmula) 221.1
Porcentaje de pureza 54 to 56% Cu
Grado Pure
Envase Frasco de vidrio
Densidad 4