missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alcohol coniferílico 99 % ACROS Organics™
Specifications
Specifications
| Nombre del producto químico o material | Coniferyl alcohol |
| Nombre de nota | 99% |
| CAS | 458-35-5 |
| Punto de fusión | De 71 °C a 76 °C |
| Color | Beige |
| Espectro infrarrojo | Auténtico |
| Intervalo de porcentaje de ensayo | 99% |
| Fórmula molecular | C10H12O3 |
| Fórmula lineal | HOC6H3(OCH3)CH=CHCH2OH |
| Número MDL | MFCD00002922 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?