Learn More
MedChemExpress Compstatin
Código de producto. 16412819
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
1 mg
Tamaño de la unidad:
1 miligramo
16412819 1 mg 1 miligramo
1 options
Código de producto. 16412819 Proveedor MedChemExpress N.º de proveedor HYP1036
MedChemExpress Compstatin, a 13-residue cyclic peptide, is a potent inhibitor of the complement system C3 with species specificity. Compstatin binds to baboon C3 and is resistant to proteolytic cleavage in baboon blood (similar to humans). Compstatin inhibits only the activation of primates’ complement system. Compstatin exhibits IC50 values of 63 μM and 12 μM for classical and alterative complement pathway, respectively.
Especificaciones
Pureza 98.34%
CAS 206645-99-0
Contenido y almacenamiento Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
Peso molecular 1550.77 g/mol
Tipo de producto Peptides
Cantidad 1 mg
Condiciones de envío Room temperature
Solubilidad H2O : 100 mg/mL (64.48 mM∣Need ultrasonic)