Learn More
Col003, MedChemExpress
Código de producto. 16427927
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
Tamaño de la unidad:
5 miligramos
16427927 5 mg 5 miligramos
1 options
Código de producto. 16427927 Proveedor Medchem Express N.º de proveedor HY124817
MedChemExpress Col003 is a selective and potent inhibitor of Hsp47 and competitively binds to the collagen binding site on Hsp47 (IC50=1.8 μM). Col003 discourages the interaction of Hsp47 with collagen and inhibits collagen secretion by destabilizing the collagen triple helix. Col003 can be used for the investigation of fibrosis
Col003 is a selective and potent inhibitor of Hsp47 and competitively binds to the collagen binding site on Hsp47 (IC50=1.8 μM). Col003 discourages the interaction of Hsp47 with collagen and inhibits collagen secretion by destabilizing the collagen triple helix. Col003 can be used for the investigation of fibrosis
Especificaciones
Nombre del producto químico o material Col003
CAS 328565-16-8
Color Yellow
Notas de grado de pureza Research
Fórmula molecular C14H11NO4
Cantidad 5 mg
Información de solubilidad DMSO : 50 mg/mL (194.37 mM; Need ultrasonic)
SMILES O=CC1=CC(CC2=CC=CC=C2)=CC([N+]([O-])=O)=C1O
Peso molecular (g/mol) 257.24
Formula Weight (peso de la fórmula) 257.24
Porcentaje de pureza 98.0%
Grado Research
Forma física Solid