Learn More
Coenzyme Q9, MedChemExpress
Código de producto. 16486077
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
1 mg
5 mg
10 mg
Tamaño de la unidad:
1 miligramo
10 miligramos
5 miligramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
16486077 5 mg 5 miligramos
16440759 1 mg 1 miligramo
16413358 10 mg 10 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 16486077 Proveedor Medchem Express N.º de proveedor HY101415
MedChemExpress Coenzyme Q9 (Ubiquinone Q9), the major form of ubiquinone in rodents, is an amphipathic molecular component of the electron transport chain that functions as an endogenous antioxidant. Coenzyme Q9 attenuates the diabetes-induced decreases in antioxidant defense mechanisms. Coenzyme Q9 improves left ventricular performance and reduces myocardial infarct size and cardiomyocyte apoptosis.
Coenzyme Q9 (Ubiquinone Q9), the major form of ubiquinone in rodents, is an amphipathic molecular component of the electron transport chain that functions as an endogenous antioxidant. Coenzyme Q9 attenuates the diabetes-induced decreases in antioxidant defense mechanisms. Coenzyme Q9 improves left ventricular performance and reduces myocardial infarct size and cardiomyocyte apoptosis.
Especificaciones
Nombre del producto químico o material Coenzyme Q9
Forma física Powder
CAS 303-97-9
Color Yellow
Cantidad 5 mg
Notas de grado de pureza Research
Fórmula molecular C54H82O4
Para utilizar con (aplicación) COVID-19-immunoregulation
Sinónimo Ubiquinone Q9 CoQ9 Ubiquinone 9
Información de solubilidad Ethanol : 33.33 mg/mL (41.91 mM; Need ultrasonic) ∣DMSO : 2 mg/mL (2.51 mM; ultrasonic and warming and heat to 80°C)
SMILES O=C1C(OC)=C(OC)C(C(C)=C1C/C=C(C)/CC/C=C(C)/CC/C=C(C)/CC/C=C(C)/CC/C=C(C)/CC/C=C(C)/CC/C=C(C)/CC/C=C(C)/CC/C=C(C)\C)=O
Peso molecular (g/mol) 795.23
Formula Weight (peso de la fórmula) 795.23
Porcentaje de pureza 98.0%