Learn More
Tocris Bioscience™ CLP 257
Descripción
CLP 257 is a selective KCC2 K+-Cl- cotransporter activator (EC50 = 616 nM). Exhibits selectivity for KCC2 over other KCC family members, NKCC1 and GABAA receptors, as well as 55 other receptors. Enhances Cl- transport and increases KCC2 plasma membrane expression in spinal slices with reduced KCC2 function. Alleviates hypersensitivity in rats with peripheral nerve injury.
Especificaciones
Especificaciones
| Nombre del producto químico o material | (5Z)-5-[(4-Fluoro-2-hydroxyphenyl)methylene]-2-(tetrahydro-1-(2H)-pyridazinyl)-4(5H)-thiazolone |
| CAS | 1181081-71-9 |
| Cantidad | 10 mg |
| Objetivo | KCC2 Activators |
| Fórmula molecular | C14H14FN3O2S |
| Formula Weight (peso de la fórmula) | 307.34 |
| Pureza | >98% |