Learn More
Clioquinol, MedChemExpress
Código de producto. 16468707
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
1 g
5 g
Tamaño de la unidad:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16468707 5 g 5 g
16411429 1 g 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16468707 Proveedor Medchem Express N.º de proveedor HY14603
MedChemExpress Clioquinol (Iodochlorhydroxyquin) is a topical antifungal agent with anticancer activity. Clioquinol acts as an oral antimicrobial agent for the research of diarrhea and skin infections. Antibiotic.
Clioquinol (Iodochlorhydroxyquin) is a topical antifungal agent with anticancer activity. Clioquinol acts as an oral antimicrobial agent for the research of diarrhea and skin infections. Antibiotic.
Especificaciones
Nombre del producto químico o material Clioquinol
Forma física Solid
CAS 130-26-7
Color Brownish Yellow
Cantidad 5 g
Notas de grado de pureza Research
Fórmula molecular C9H5ClINO
Para utilizar con (aplicación) COVID-19-immunoregulation
Sinónimo Iodochlorhydroxyquin
Información de solubilidad DMSO : ≥ 53.33 mg/mL (174.57 mM)
SMILES ClC1=C(C=CC=N2)C2=C(O)C(I)=C1
Peso molecular (g/mol) 305.5
Formula Weight (peso de la fórmula) 305.5
Porcentaje de pureza 98.04%