missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CKS2 Recombinant Rabbit Monoclonal Antibody (K01_3K53)
GREENER_CHOICE

Código de producto. 30829995
Change view
Click to view available options
Cantidad:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad
30829995 100 μL
30828101 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 30829995 Proveedor Invitrogen™ N.º de proveedor MA564359

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Recombinant Monoclonal Antibody

CKS2 Recombinant Monoclonal Antibody for Western Blot

CKS2 protein binds to the catalytic subunit of the cyclin dependent kinases and is essential for their biological function. The CKS2 mRNA is found to be expressed in different patterns through the cell cycle in HeLa cells, which reflects specialized role for the encoded protein. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antígeno CKS2
Aplicaciones Western Blot
Clasificación Recombinant Monoclonal
Clon K01_3K53
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación 50mM tris glycine with 40% glycerol, 0.05% BSA, 150mM NaCl and 0.01% sodium azide; pH 7.4
génica Cks2
N.º de referencia del gen P33552, P56390
Alias de gen 1110038L14Rik; CDC28 protein kinase 2; CDC28 protein kinase regulatory subunit 2; CKS1(S. cerevisiae Cdc28/Cdc2 kinase subunit) homolog-2; CKS2; CKS-2; CKSHS2; Cyclin-dependent kinases regulatory subunit 2; RGD1562047; RP11-8C21.2
Símbolos de los genes Cks2
Especie del huésped Rabbit
Inmunógeno Peptide.
Método de purificación Protein A/G
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 1164, 498709, 66197
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.