Learn More
Tocris Bioscience™ cis-22a
Descripción
cis-22a is a TRPV6 inhibitor (IC50 = 0.32 μM), which exhibits selectivity against related TRPV channels and calcium channels. cis-22a displays antiproliferative effects on T47D human breast cancer cells.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 1-[cis-4-(3-Methylphenyl)cyclohexyl]-4-(3-pyridinyl)piperazine ditrifluoroacetate |
| CAS | 1819366-84-1 |
| Cantidad | 10 mg |
| Objetivo | TRPV Inhibitors |
| Fórmula molecular | C22H29N30.2 CF3CO2H |
| Pureza | 0.98 |