Learn More
Tocris Bioscience™ Cinacalcet hydrochloride
Low molecular weight, organic compound that may help regulate biological process.
Marca: Tocris Bioscience™ 6170/10
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Cinacalcet hydrochloride is a calcium-sensing receptor (CaSR) allosteric agonist. Activates CaSR signaling in cellular proliferation and phosphatidylinositol (PI) hydrolysis assays. Reduces serum parathyroid hormone levels in rats. Also potent CYP2D2 inhibitor (IC50 = 87 nM) and L-type calcium channel blocker. Orally bioavailable.Especificaciones
| Calcium-sensing Receptor (CaSR) Allosteric Agonist, also CYP2D2 Inhibitor and Ca2+ Channel Blocker | |
| 364782-34-3 | |
| 393.87g/mol | |
| Calcium-Sensing Receptor Agonists |
| Calcium-Sensing Receptor | |
| C22H22F3N.HCl | |
| Store at -20°C | |
| 10 mg |
Título del producto
For Research Use Only