Learn More
Tocris Bioscience™ CID 16020046
Descripción
CID 16020046 is a selective GPR55 antagonist. Inhibits LPI-induced Ca2+ signaling (IC50 = 0.21 μM in HEK-GPR55 cells), ERK1/2 phosphorylation and GPR55-mediated transcription factor activation. Displays weak inhibition of acetylcholinesterase, μ-opioid receptor, KCNH2 and hERG. Decreases LPI-induced GPR55 internalization. Reduces experimental intestinal inflammation in mice.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 4-[4,6-Dihydro-4-(3-hydroxyphenyl)-3-(4-methylphenyl)-6-oxopyrrolo[3,4-c]pyrazol-5(1H)-yl]benzoic acid |
| CAS | 834903-43-4 |
| Intervalo de porcentaje de ensayo | >99% |
| Cantidad | 25 mg |
| Fórmula molecular | C25H19N3O4 |
| Formula Weight (peso de la fórmula) | Observed MW: 425.44 |