Learn More
Ciclopirox, Tocris Bioscience™
Pan-histone demethylase inhibitor
Marca: Tocris Bioscience 6384/10
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Ciclopirox is a pan-histone demethylase inhibitor. Exhibits no significant effect on histone deacetylases. Inhibits MYC-signaling and proliferation of neuroblastoma cells. Promotes Cdc25A-degradation in cancer cells. Antifungal.Especificaciones
| 6-Cyclohexyl-1-hydroxy-4-methyl-2(1H)-pyridinone | |
| 29342-05-0 | |
| >99% | |
| 10 mg |