Learn More
R&D Systems™ ci-IP3/PM
Description
ci-IP3/PM is a caged inositol triphosphate. Cell permeable. UV photolysis (λ = 330 nm) in HeLa cells releases i-IP3, promoting Ca2+-release from internal stores.
F 127 (Cat. No. 6253) for the solubilization of ci-IP3/PM is also available.
Specifications
Specifications
| CAS | 1009832-82-9 |
| Cantidad | 10 μg |
| Línea de productos | Tocris |
| Fórmula molecular | C42H64NO31P3 |
| Formula Weight (peso de la fórmula) | 1171.87g/mol |
| Tipo de producto | Inositol Triphosphate |
Para uso exclusivo en investigación