Learn More
Thermo Scientific Chemicals Cromazurol B
Código de producto. 16594264 Tienda Thermo Scientific Chemicals Productos
Cambiar vista
Click to view available options
Cantidad:
5 g
25 g
Tamaño de la unidad:
25 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16594264 5 g 5 g
16504274 25 g 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16594264 Proveedor Thermo Scientific Chemicals N.º de proveedor 464530050
Solicitar formato a granel o personalizado
CAS: 1796-92-5 | C23H14Cl2Na2O6 | 503.24 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
CAS 1796-92-5
Fórmula molecular C23H14Cl2Na2O6
Peso molecular (g/mol) 503.24
Clave InChI JROAZQFKSSYEBL-YWABOPJLSA-L
Nombre IUPAC disodio 5-{[(1E)-3-carboxilato-5-metil-4-oxiclohexa-2,5-dien-1-ilideno](2,6-diclorofenil)metil}-2-hidroxi-3-metilbenzoato
SMILES [Na+].[Na+].CC1=CC(=CC(C([O-])=O)=C1O)C(=C1\C=C(C)C(=O)C(=C1)C([O-])=O)\C1=C(Cl)C=CC=C1Cl
Especificaciones
Nombre del producto químico o material Chromazurol B
Cantidad 5 g
Formula Weight (peso de la fórmula) 503.25