Learn More
Thermo Scientific Chemicals Clorhidrato de cloroprocaína
Código de producto. 16514484 Tienda Thermo Scientific Chemicals Productos
Cambiar vista
Click to view available options
Cantidad:
250 mg
1 g
Tamaño de la unidad:
1 g
250 miligramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16514484 250 mg 250 miligramos
16504484 1 g 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16514484 Proveedor Thermo Scientific Chemicals N.º de proveedor 465372500
Solicitar formato a granel o personalizado
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
CAS 3858-89-7
Fórmula molecular C13H20Cl2N2O2
Peso molecular (g/mol) 307.22
Clave InChI SZKQYDBPUCZLRX-UHFFFAOYSA-N
Nombre IUPAC clorhidrato de 2-(dietilamino)etilo 4-amino-2-clorobenzoato
SMILES Cl.CCN(CC)CCOC(=O)C1=CC=C(N)C=C1Cl
Especificaciones
Nombre del producto químico o material Chloroprocaine hydrochloride
Punto de fusión 173.0°C to 178.0°C
Cantidad 250 mg
Información de solubilidad Solubility aq. soln.: soluble.
Formula Weight (peso de la fórmula) 307.22